Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hacd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199861.1 | NP_956155.1 | 4 - 5 | 414 | 66 | |
GenBank mRNA | BC044396.1 | - | 4 - 5 | 414 | 66 | |
BC165084.1 | - | 4 - 5 | 376 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |