Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hagh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200043.2 | NP_956337.1 | 4 - 5 | 442 | 108 | |
XM_005163865.4 | XP_005163922.1 | 4 - 5 | 506 | 108 | ||
XM_009299331.3 | XP_009297606.1 | 4 - 5 | 506 | 108 | ||
GenBank mRNA | AY398356.1 | - | 4 - 5 | 442 | 108 | |
BC060913.1 | - | 3 - 4 | 330 | 108 | ||
BC066607.1 | - | 4 - 5 | 428 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |