Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lmo4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212689.2 | NP_997854.2 | 4 - 5 | 866 | 89 | |
GenBank mRNA | AF398515.2 | - | 4 - 5 | 822 | 89 | |
BC076053.1 | - | 4 - 5 | 840 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |