Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: esrrga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212954.1 | NP_998119.1 | 3 - 4 | 635 | 87 | |
XM_009293126.3 | XP_009291401.1 | 5 - 6 | 1098 | 87 | ||
XM_021467130.1 | XP_021322805.1 | 5 - 6 | 1111 | 87 | ||
GenBank mRNA | AY556397.1 | - | 3 - 4 | 635 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |