Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gmppaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002196.1 | NP_001002196.1 | 4 - 5 | 574 | 100 | |
XM_005167476.4 | XP_005167533.1 | 4 - 5 | 705 | 100 | ||
GenBank mRNA | BC074036.1 | - | 4 - 5 | 574 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |