Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cldn7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002340.1 | NP_001002340.1 | 1 - 2 | 420 | 98 | |
GenBank mRNA | BC075926.1 | - | 1 - 2 | 420 | 98 | |
BC153583.1 | - | 1 - 2 | 339 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |