Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nck2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003492.2 | NP_001003492.2 | 2 - 3 | 421 | 88 | |
GenBank mRNA | BC078356.1 | - | 2 - 3 | 389 | 88 | |
BC096988.1 | - | 2 - 3 | 421 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |