Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003889.1 | NP_001003889.1 | 2 - 3 | 252 | 58 | |
GenBank mRNA | AY648806.1 | - | 2 - 3 | 252 | 58 | |
BC085580.1 | - | 2 - 3 | 247 | 58 | ||
BC165077.1 | - | 2 - 3 | 242 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |