Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudt22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006069.1 | NP_001006069.1 | 6 - 7 | 986 | 97 | |
XM_005173118.4 | XP_005173175.1 | 6 - 7 | 1054 | 97 | ||
XM_021481006.1 | XP_021336681.1 | 5 - 6 | 1182 | 97 | ||
GenBank mRNA | BC083253.1 | - | 6 - 7 | 986 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |