Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: six1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009904.1 | NP_001009904.1 | 1 - 2 | 798 | 92 | |
XM_021480569.1 | XP_021336244.1 | 2 - 3 | 1099 | 92 | ||
GenBank mRNA | BC076015.1 | - | 1 - 2 | 798 | 92 | |
EU109509.1 | - | 1 - 2 | 564 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |