Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: angptl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014820.1 | NP_001014820.1 | 3 - 4 | 1254 | 67 | |
XM_005160416.4 | XP_005160473.1 | 4 - 5 | 1721 | 67 | ||
GenBank mRNA | AB199647.1 | - | 3 - 4 | 1254 | 67 | |
BC151871.1 | - | 4 - 5 | 1639 | 67 | ||
BC163080.1 | - | 3 - 4 | 1296 | 67 | ||
BC163101.1 | - | 3 - 4 | 1296 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |