Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhof | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020642.1 | NP_001018478.1 | 4 - 5 | 644 | 63 | |
GenBank mRNA | BC095646.1 | - | 4 - 5 | 644 | 63 | |
BC165017.1 | - | 4 - 5 | 496 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |