Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: il22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020792.1 | NP_001018628.1 | 1 - 2 | 272 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons |