Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: illr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040040.1 | NP_001035129.1 | 2 - 3 | 196 | 116 | |
GenBank mRNA | AY986758.1 | - | 2 - 3 | 196 | 116 | |
BC124080.1 | - | 2 - 3 | 222 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |