Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mov10b.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044342.2 | NP_001037807.2 | 14 - 15 | 2192 | 101 | |
GenBank mRNA | BC171375.1 | - | 14 - 15 | 2192 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |