Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spata2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080191.2 | NP_001073660.2 | 2 - 3 | 979 | 88 | |
XM_005161908.4 | XP_005161965.1 | 2 - 3 | 912 | 88 | ||
XM_005161909.4 | XP_005161966.1 | 2 - 3 | 1010 | 88 | ||
GenBank mRNA | BC150216.1 | - | 2 - 3 | 979 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |