Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oxsr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098747.1 | NP_001092217.1 | 9 - 10 | 1030 | 81 | |
GenBank mRNA | BC124256.1 | - | 9 - 10 | 1077 | 81 | |
BC142933.1 | - | 9 - 10 | 1030 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |