Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105594.1 | NP_001099064.1 | 2 - 3 | 887 | 75 | |
GenBank mRNA | BC142873.1 | - | 2 - 3 | 653 | 75 | |
BC153383.1 | - | 2 - 3 | 887 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |