Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105594.1 | NP_001099064.1 | 4 - 5 | 1174 | 88 | |
GenBank mRNA | BC133172.1 | - | 4 - 5 | 943 | 88 | |
BC142873.1 | - | 4 - 5 | 940 | 88 | ||
BC153383.1 | - | 4 - 5 | 1174 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |