Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gspt1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198806.1 | NP_942101.1 | 11 - 12 | 1569 | 77 | |
GenBank mRNA | BC053244.1 | - | 11 - 12 | 1569 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |