Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201297.1 | NP_958454.1 | 2 - 3 | 132 | 59 | |
XM_005163797.4 | XP_005163854.1 | 2 - 3 | 217 | 59 | ||
GenBank mRNA | BC055384.1 | - | 2 - 3 | 132 | 59 | |
BC065895.1 | - | 2 - 3 | 133 | 59 | ||
KM655719.1 | - | 1 - 2 | 88 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |