Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205553.1 | NP_991116.1 | 2 - 3 | 80 | ||
NM_205766.1 | NP_991329.1 | 2 - 3 | 367 | 80 | ||
GenBank mRNA | BC044545.1 | - | 2 - 3 | 367 | 80 | |
BC065643.1 | - | 2 - 3 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |