Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prpf38b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213489.1 | NP_998654.1 | 1 - 2 | 411 | 127 | |
XM_005171371.4 | XP_005171428.1 | 1 - 2 | 431 | 127 | ||
XM_005171372.4 | XP_005171429.1 | 1 - 2 | 431 | 127 | ||
XM_009298770.3 | XP_009297045.1 | 1 - 2 | 432 | 127 | ||
GenBank mRNA | BC061448.1 | - | 1 - 2 | 411 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |