Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbm42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003850.1 | NP_001003850.1 | 10 - 11 | 1273 | 82 | |
GenBank mRNA | AY648798.1 | - | 10 - 11 | 1273 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Danio rerio Probe spans exons |