Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dmrt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005779.2 | NP_001005779.2 | 1 - 2 | 585 | 70 | |
GenBank mRNA | BC083273.1 | - | 1 - 2 | 585 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |