Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fli1rs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008780.1 | NP_001008780.1 | 3 - 4 | 499 | 60 | |
XM_005158200.3 | XP_005158257.1 | 3 - 4 | 575 | 60 | ||
XM_005158202.4 | XP_005158259.1 | 3 - 4 | 491 | 60 | ||
XM_005158203.4 | XP_005158260.1 | 3 - 4 | 491 | 60 | ||
XM_005158205.4 | XP_005158262.1 | 3 - 4 | 491 | 60 | ||
XM_009292528.3 | XP_009290803.1 | 3 - 4 | 607 | 60 | ||
XM_009292529.3 | XP_009290804.1 | 3 - 4 | 607 | 60 | ||
XM_009292530.3 | XP_009290805.1 | 3 - 4 | 606 | 60 | ||
XM_009292531.3 | XP_009290806.1 | 3 - 4 | 606 | 60 | ||
GenBank mRNA | AY839950.1 | - | 3 - 4 | 499 | 60 | |
BC055627.1 | - | 3 - 4 | 423 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |