Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smyd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013550.1 | NP_001013568.1 | 3 - 4 | 494 | 126 | |
GenBank mRNA | BC091465.1 | - | 3 - 4 | 494 | 126 | |
DQ851823.1 | - | 3 - 4 | 377 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |