Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc38a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017644.1 | NP_001017644.1 | 7 - 8 | 699 | 89 | |
GenBank mRNA | BC092805.1 | - | 7 - 8 | 699 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |