Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020630.1 | NP_001018466.1 | 6 - 7 | 935 | 105 | |
XM_021480399.1 | XP_021336074.1 | 6 - 7 | 947 | 105 | ||
GenBank mRNA | BC095608.1 | - | 6 - 7 | 935 | 105 | |
BC165429.1 | - | 5 - 6 | 638 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |