Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scn1lab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ149504.1 | - | 13 - 14 | 2333 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |