Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045105.1 | NP_001038570.1 | 4 - 5 | 1639 | 86 | |
XM_017357352.2 | XP_017212841.1 | 4 - 5 | 1947 | 86 | ||
XM_021478287.1 | XP_021333962.1 | 4 - 5 | 1688 | 86 | ||
GenBank mRNA | BC134845.1 | - | 4 - 5 | 1550 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |