Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045105.1 | NP_001038570.1 | 5 - 6 | 1735 | 110 | |
XM_017357352.2 | XP_017212841.1 | 5 - 6 | 2043 | 110 | ||
GenBank mRNA | BC134845.1 | - | 5 - 6 | 1646 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |