Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adpgk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079965.2 | NP_001073434.1 | 5 - 6 | 801 | 61 | |
XM_005162956.4 | XP_005163013.1 | 6 - 7 | 940 | 61 | ||
XM_005162957.4 | XP_005163014.1 | 5 - 6 | 1017 | 61 | ||
GenBank mRNA | BC125924.1 | - | 5 - 6 | 735 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |