Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mylipb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080022.2 | NP_001073491.1 | 2 - 3 | 291 | 85 | |
GenBank mRNA | BC124450.1 | - | 2 - 3 | 292 | 85 | |
BC165740.1 | - | 2 - 3 | 308 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |