Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gata2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131233.1 | NP_571308.1 | 5 - 6 | 1440 | 64 | |
XM_009305781.3 | XP_009304056.1 | 5 - 6 | 1520 | 64 | ||
XM_009305782.3 | XP_009304057.1 | 5 - 6 | 1657 | 64 | ||
XM_021479548.1 | XP_021335223.1 | 4 - 5 | 1122 | 64 | ||
GenBank mRNA | BC053131.1 | - | 5 - 6 | 1440 | 64 | |
U18312.1 | - | 2 - 3 | 289 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |