Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc4a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198338.1 | NP_938152.1 | 4 - 5 | 291 | 78 | |
GenBank mRNA | AF350072.1 | - | 4 - 5 | 291 | 78 | |
AY098936.1 | - | 4 - 5 | 324 | 78 | ||
BC163932.1 | - | 4 - 5 | 277 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |