Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grhprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205685.2 | NP_991248.2 | 7 - 8 | 1046 | 99 | |
XM_005168340.4 | XP_005168397.1 | 7 - 8 | 1011 | 99 | ||
GenBank mRNA | BC065431.1 | - | 7 - 8 | 906 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |