Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pnpla3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002338.1 | NP_001002338.1 | 8 - 9 | 1178 | 70 | |
GenBank mRNA | BC075928.1 | - | 8 - 9 | 1178 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |