Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hvcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002346.1 | NP_001002346.1 | 4 - 5 | 607 | 73 | |
GenBank mRNA | BC075916.1 | - | 4 - 5 | 607 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |