Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: med31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002417.2 | NP_001002417.2 | 2 - 3 | 240 | 83 | |
GenBank mRNA | BC076157.1 | - | 2 - 3 | 235 | 83 | |
EH553004.1 | - | 2 - 3 | 279 | 83 | ||
EH600668.1 | - | 2 - 3 | 630 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |