Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: synpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004532.2 | NP_001004532.1 | 2 - 3 | 232 | 75 | |
GenBank mRNA | BC086853.1 | - | 2 - 3 | 232 | 75 | |
BC164641.1 | - | 2 - 3 | 179 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |