Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcyt1bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037374.1 | NP_001032451.1 | 5 - 6 | 615 | 72 | |
XM_005157481.4 | XP_005157538.1 | 5 - 6 | 655 | 72 | ||
GenBank mRNA | BC109442.1 | - | 5 - 6 | 650 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |