Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sycp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040350.1 | NP_001035440.1 | 6 - 7 | 735 | 63 | |
GenBank mRNA | BC115343.1 | - | 6 - 7 | 735 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |