Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043321.1 | NP_001036786.1 | 1 - 2 | 232 | 55 | |
GenBank mRNA | AB298722.1 | - | 1 - 2 | 129 | 55 | |
BC049029.1 | - | 1 - 2 | 221 | 55 | ||
BC171420.1 | - | 1 - 2 | 220 | 55 | ||
BC171422.1 | - | 1 - 2 | 220 | 55 | ||
DQ639953.1 | - | 1 - 2 | 232 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |