Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077385.2 | NP_001070853.1 | 2 - 3 | 751 | 107 | |
GenBank mRNA | BC124692.1 | - | 2 - 3 | 754 | 107 | |
BC165550.1 | - | 2 - 3 | 708 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |