Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sult2st3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078168.2 | NP_001071636.2 | 5 - 6 | 800 | 69 | |
XM_017351367.1 | XP_017206856.1 | 6 - 7 | 871 | 69 | ||
GenBank mRNA | BC115350.1 | - | 5 - 6 | 800 | 69 | |
DQ640388.1 | - | 5 - 6 | 754 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |