Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacng6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083091.1 | NP_001076560.1 | 3 - 4 | 613 | 73 | |
XM_005159405.4 | XP_005159462.1 | 4 - 5 | 1550 | 73 | ||
XM_009294017.3 | XP_009292292.1 | 5 - 6 | 1359 | 73 | ||
XM_009294018.3 | XP_009292293.1 | 4 - 5 | 1256 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Danio rerio Probe spans exons |