Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hspb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100958.1 | NP_001094428.1 | 1 - 2 | 435 | 95 | |
GenBank mRNA | EF614998.1 | - | 1 - 2 | 435 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Danio rerio Probe spans exons |