Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131555.2 | NP_571630.2 | 6 - 7 | 1284 | 78 | |
GenBank mRNA | AF280090.1 | - | 6 - 7 | 1223 | 78 | |
BC129260.1 | - | 6 - 7 | 1284 | 78 | ||
BC164222.1 | - | 6 - 7 | 1205 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |