Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2ad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152954.1 | NP_694486.1 | 3 - 4 | 529 | 91 | |
GenBank mRNA | AF248042.2 | - | 3 - 4 | 528 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |